Learn More
Eurofins Discoverx PathHunter Human PD-1 (SHP2) Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455951
Encompass
The PathHunter Human PD-1 (SHP2) Signaling Stable Cell Line Assay (Jurkat) measures PD-1 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55951 Supplier Eurofins Discoverx Supplier No. 7931106C19
The PathHunter Human PD-1 (SHP2) Signaling Stable Cell Line Assay (Jurkat) measures PD-1 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.