Learn More
Eurofins Discoverx PathHunter Human LEPR/LEPR Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456058
Encompass
The PathHunter Human LEPR/LEPR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of LEPR/LEPR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56058 Supplier Eurofins Discoverx Supplier No. 7931107C3
The PathHunter Human LEPR/LEPR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of LEPR/LEPR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.