Learn More
Eurofins Discoverx PathHunter Human TPOR/TPOR Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456059
Encompass
The PathHunter Human TPOR/TPOR Dimerization Stable Cell Line Assay (HEK 293) measures ligand-induced dimerization of TPOR/TPOR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56059 Supplier Eurofins Discoverx Supplier No. 7931108C1
The PathHunter Human TPOR/TPOR Dimerization Stable Cell Line Assay (HEK 293) measures ligand-induced dimerization of TPOR/TPOR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.