Learn More
Eurofins Discoverx PathHunter Human BTLA Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456167
Encompass
The PathHunter Human BTLA (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures BTLA signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56167 Supplier Eurofins Discoverx Supplier No. 7931112C19
The PathHunter Human BTLA (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures BTLA signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.