Learn More
Eurofins Discoverx PathHunter Human FGFR1-alpha-Klotho Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455851
Encompass
The PathHunter Human FGFR1-alpha-Klotho Stable Cell Line Functional Kinase Assay (U2OS) measures FGFR1-α-Klotho receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55851 Supplier Eurofins Discoverx Supplier No. 7931117C3
The PathHunter Human FGFR1-alpha-Klotho Stable Cell Line Functional Kinase Assay (U2OS) measures FGFR1-α-Klotho receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.