Learn More
Eurofins Discoverx PathHunter Human TORC3 (CRTC3) Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000456061
Encompass
The PathHunter Human TORC3 (CRTC3) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure CRTC/CREB pathway activation via nuclear translocation of TORC3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56061 Supplier Eurofins Discoverx Supplier No. 7931120C3
The PathHunter Human TORC3 (CRTC3) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure CRTC/CREB pathway activation via nuclear translocation of TORC3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.