Learn More
Eurofins Discoverx PathHunter Human IkB Degradation Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456024
Encompass
The PathHunter Human IkB Degradation Stable Cell Line Assay (Jurkat) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56024 Supplier Eurofins Discoverx Supplier No. 7931121C19
The PathHunter Human IkB Degradation Stable Cell Line Assay (Jurkat) measures pathway activation-induced degradation of IκB target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.