Learn More
Eurofins Discoverx PathHunter Human Heregulin β1 (EGFR/ErbB3) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456005
Encompass
The PathHunter Human Heregulin β1 (EGFR/ErbB3) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of EGFR/ErbB3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56005 Supplier Eurofins Discoverx Supplier No. 7931125C3
The PathHunter Human Heregulin β1 (EGFR/ErbB3) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of EGFR/ErbB3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.