Learn More
Eurofins Discoverx PathHunter Human TGF-β1 (TGFBR1/TGFBR2/ENG) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456062
Encompass
The PathHunter Human TGF-β1 (TGFBR1/TGFBR2/ENG) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of TGFBR1/TGFBR2 when co-expressed with Endoglin (ENG). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56062 Supplier Eurofins Discoverx Supplier No. 7931122C3
The PathHunter Human TGF-β1 (TGFBR1/TGFBR2/ENG) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of TGFBR1/TGFBR2 when co-expressed with Endoglin (ENG). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.