Learn More
Eurofins Discoverx PathHunter Human CD28 Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456004
Encompass
The PathHunter Human CD28 (GRB2) Signaling Stable Cell Line Assay (Jurkat) measures CD28 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56004 Supplier Eurofins Discoverx Supplier No. 7931126C19
The PathHunter Human CD28 (GRB2) Signaling Stable Cell Line Assay (Jurkat) measures CD28 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.