Learn More
Eurofins Discoverx PathHunter Human ICOS Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455507
Encompass
The PathHunter Human ICOS (P85) Signaling Stable Cell Line Assay (Jurkat) measures ICOS signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55507 Supplier Eurofins Discoverx Supplier No. 7931127C19
The PathHunter Human ICOS (P85) Signaling Stable Cell Line Assay (Jurkat) measures ICOS signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.