Learn More
Eurofins Discoverx PathHunter Human PD-L1/PD-L1 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455854
Encompass
The PathHunter Human PD-L1/PD-L1 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of PD-L1/PD-L1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55854 Supplier Eurofins Discoverx Supplier No. 7931129C3
The PathHunter Human PD-L1/PD-L1 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of PD-L1/PD-L1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.