Learn More
Eurofins Discoverx PathHunter Human IL-18 (IL18R1/IL18RAP) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455990
Encompass
The PathHunter Human IL-18 (IL18R1/IL18RAP) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL18R1/IL18RAP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55990 Supplier Eurofins Discoverx Supplier No. 7931131C3
The PathHunter Human IL-18 (IL18R1/IL18RAP) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL18R1/IL18RAP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.