Learn More
Eurofins Discoverx PathHunter Human PD-1/PD-1 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456141
Encompass
The PathHunter Human PD-1/PD-1 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of PD-1/PD-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56141 Supplier Eurofins Discoverx Supplier No. 7931130C3
The PathHunter Human PD-1/PD-1 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of PD-1/PD-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.