Learn More
Eurofins Discoverx PathHunter Human OPRK1 (Opioid Kappa) β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000456016
Encompass
The PathHunter Human OPRK1 (Opioid Kappa) β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure OPRK1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56016 Supplier Eurofins Discoverx Supplier No. 7930234C3
The PathHunter Human OPRK1 (Opioid Kappa) β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure OPRK1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.