Learn More
Eurofins Discoverx PathHunter Human T-Cell Receptor Activation Pathway Reporter Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455853
Encompass
The PathHunter Human T-Cell Receptor Activation Pathway NFAT Reporter Stable Cell Line Assay (Jurkat) measures induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55853 Supplier Eurofins Discoverx Supplier No. 7931141C19
The PathHunter Human T-Cell Receptor Activation Pathway NFAT Reporter Stable Cell Line Assay (Jurkat) measures induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.