Learn More
Eurofins Discoverx PathHunter Human STAT3 Pathway Reporter Cell-Based Assay Kit (HepG2)
Catalog No. 5000456168
Encompass
The PathHunter Human STAT3 Pathway Reporter Stable Cell Line Assay (HepG2) measures induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56168 Supplier Eurofins Discoverx Supplier No. 7931144C44
The PathHunter Human STAT3 Pathway Reporter Stable Cell Line Assay (HepG2) measures induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.