Learn More
Eurofins Discoverx PathHunter Human BCMA Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455988
Encompass
The PathHunter Human BCMA Stable Cell Line Internalization Assay (U2OS) measures internalization of the BCMA receptor via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55988 Supplier Eurofins Discoverx Supplier No. 7931145C3
The PathHunter Human BCMA Stable Cell Line Internalization Assay (U2OS) measures internalization of the BCMA receptor via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.