Learn More
Eurofins Discoverx PathHunter Human CD33 (SIGLEC3) Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456140
Encompass
The PathHunter Human CD33 (SIGLEC3) Stable Cell Line Internalization Assay (U2OS) measures internalization of the CD33 receptor via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56140 Supplier Eurofins Discoverx Supplier No. 7931148C3
The PathHunter Human CD33 (SIGLEC3) Stable Cell Line Internalization Assay (U2OS) measures internalization of the CD33 receptor via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.