Learn More
Eurofins Discoverx PathHunter Human IL-17E (IL17RA/IL17RB) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000456053
Encompass
The PathHunter Human IL-17E (IL17RA/IL17RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL17RA and IL17RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56053 Supplier Eurofins Discoverx Supplier No. 7931150C3
The PathHunter Human IL-17E (IL17RA/IL17RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL17RA and IL17RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.