Learn More
Eurofins Discoverx PathHunter Human Mer Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000456054
Encompass
The PathHunter Human Mer Stable Cell Line Functional Kinase Assay (U2OS) measures activation through receptor dimerization leading to SH2-binding protein recruitment. This complete kit includes cell line vials and all required reagents for cell culture, maintenance and for running the assay.
Catalog No. 50-004-56054 Supplier Eurofins Discoverx Supplier No. 7931151C3
The PathHunter Human Mer Stable Cell Line Functional Kinase Assay (U2OS) measures activation through receptor dimerization leading to SH2-binding protein recruitment. This complete kit includes cell line vials and all required reagents for cell culture, maintenance and for running the assay.