Learn More
Eurofins Discoverx PathHunter Human RANKL (RANK) Signaling Reporter Cell-Based Assay Kit (U2OS)
Catalog No. 5000455992
Encompass
The PathHunter Human RANKL (RANK) Signaling NF-kB Reporter Stable Cell Line Assay (U2OS) measures NF-kB pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55992 Supplier Eurofins Discoverx Supplier No. 7931156C3
The PathHunter Human RANKL (RANK) Signaling NF-kB Reporter Stable Cell Line Assay (U2OS) measures NF-kB pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.