Learn More
Eurofins Discoverx PathHunter Mouse PD-1 Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456108
Encompass
The PathHunter Mouse PD-1 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures PD-1 signal by recruitment of SH2 domain protein to the activated receptor. Please note this product expresses the mouse version of PD-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56108 Supplier Eurofins Discoverx Supplier No. 7931158C19
The PathHunter Mouse PD-1 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures PD-1 signal by recruitment of SH2 domain protein to the activated receptor. Please note this product expresses the mouse version of PD-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.