Learn More
Eurofins Discoverx PathHunter Human FGFR3 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455460
Encompass
The PathHunter Human FGFR3 Stable Cell Line Functional Kinase Assay (U2OS) measures activation via receptor dimerization leading to SH2-binding protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55460 Supplier Eurofins Discoverx Supplier No. 7931160C3
The PathHunter Human FGFR3 Stable Cell Line Functional Kinase Assay (U2OS) measures activation via receptor dimerization leading to SH2-binding protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.