Learn More
Eurofins Discoverx PathHunter Mouse CTLA4 Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455461
Encompass
The PathHunter Mouse CTLA4 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures CTLA4 signal by recruitment of SH2 domain protein to the activated receptor. Please note this product expresses the mouse version of CTLA-4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55461 Supplier Eurofins Discoverx Supplier No. 7931161C19
Add to Cart
Add to Cart
The PathHunter Mouse CTLA4 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures CTLA4 signal by recruitment of SH2 domain protein to the activated receptor. Please note this product expresses the mouse version of CTLA-4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.