Learn More
Eurofins Discoverx PathHunter Human NF-kB Pathway Reporter Cell-Based Assay Kit (HEK 293)
Catalog No. 5000456064
Encompass
The PathHunter NF-kB Pathway Reporter Stable Cell Line Assay (HEK 293) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology.  This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56064 Supplier Eurofins Discoverx Supplier No. 7931164C1
Add to Cart
Add to Cart
The PathHunter NF-kB Pathway Reporter Stable Cell Line Assay (HEK 293) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology.  This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.