Learn More
Eurofins Discoverx PathHunter Human IKZF3v1 Degradation Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455462
Encompass
The PathHunter Human IKZF3v1 Degradation Stable Cell Line Assay (HEK 293) measures pathway activation-induced degradation of IKZF3v1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55462 Supplier Eurofins Discoverx Supplier No. 7931165C1
The PathHunter Human IKZF3v1 Degradation Stable Cell Line Assay (HEK 293) measures pathway activation-induced degradation of IKZF3v1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.