Learn More
Eurofins Discoverx PathHunter Human CD27 Signaling Reporter Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455991
Encompass
The PathHunter Human CD27 Signaling NF-kB Reporter Stable Cell Line Assay (HEK 293) measures NF-kB pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55991 Supplier Eurofins Discoverx Supplier No. 7931166C1
The PathHunter Human CD27 Signaling NF-kB Reporter Stable Cell Line Assay (HEK 293) measures NF-kB pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.