Learn More
Eurofins Discoverx PathHunter Human TREM1-DAP12 Signaling Reporter Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456063
Encompass
The PathHunter Human TREM1-DAP12 Signaling NFAT Reporter Stable Cell Line Assay (Jurkat) monitors the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56063 Supplier Eurofins Discoverx Supplier No. 7931167C19
The PathHunter Human TREM1-DAP12 Signaling NFAT Reporter Stable Cell Line Assay (Jurkat) monitors the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.