Learn More
Eurofins Discoverx PathHunter Human IL-11 (IL11RA) Signaling Reporter Cell-Based Assay Kit (HepG2)
Catalog No. 5000456026
Encompass
The PathHunter Human IL-11 (IL11RA) Signaling STAT3 Reporter Stable Cell Line Assay (HepG2) measures STAT3 pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56026 Supplier Eurofins Discoverx Supplier No. 7931168C044
The PathHunter Human IL-11 (IL11RA) Signaling STAT3 Reporter Stable Cell Line Assay (HepG2) measures STAT3 pathway activation through detection of induced expression levels of a tagged reporter protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.