Learn More
Eurofins Discoverx PathHunter Human LAIR1 Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455499
Encompass
The PathHunter Human LAIR1 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures LAIR1 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55499 Supplier Eurofins Discoverx Supplier No. 7931170C19
The PathHunter Human LAIR1 (SHP1) Signaling Stable Cell Line Assay (Jurkat) measures LAIR1 signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.