Learn More
Eurofins Discoverx PathHunter Human GM-CSF Signaling Reporter Cell-Based Assay Kit (U937)
Catalog No. 5000456142
Encompass
The PathHunter Human GM-CSF Signaling STAT5 Reporter Stable Cell Line Assay (U937) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56142 Supplier Eurofins Discoverx Supplier No. 7931171C054
The PathHunter Human GM-CSF Signaling STAT5 Reporter Stable Cell Line Assay (U937) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.