Learn More
Eurofins Discoverx PathHunter Human IL-7 (IL7R/IL2RG) Signaling Reporter Cell-Based Assay Kit (REH)
Supplier:  Eurofins Discoverx 7931173C055
Encompass
The PathHunter Human IL-7 (IL7R/IL2RG) Signaling STAT5 Reporter Stable Cell Line Assay (REH) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56027