Learn More
Eurofins Discoverx PathHunter Human Signaling Reporter Cell-Based Assay Kit (HEK293)
Catalog No. 5000456087
Encompass
The PathHunter Human IFN gamma Signaling IRF1/GAS Reporter Stable Cell Line Assay (HEK293) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56087 Supplier Eurofins Discoverx Supplier No. 7931174C1
The PathHunter Human IFN gamma Signaling IRF1/GAS Reporter Stable Cell Line Assay (HEK293) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.