Learn More
Eurofins Discoverx PathHunter Human SREBP1c Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000456086
Encompass
The PathHunter Human SREBP1c Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure effects on cholesterol transport via detecting SREBP1c nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56086 Supplier Eurofins Discoverx Supplier No. 7931175C3
The PathHunter Human SREBP1c Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure effects on cholesterol transport via detecting SREBP1c nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.