Learn More
Eurofins Discoverx PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Signaling Reporter Cell-Based Assay Kit (REH)
Catalog No. 5000455971
Encompass
The PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Signaling STAT5 Reporter Stable Cell Line Assay (REH) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55971 Supplier Eurofins Discoverx Supplier No. 7931178C055
The PathHunter Human IL-15 (IL-15R (IL2RB/IL2RG)) Signaling STAT5 Reporter Stable Cell Line Assay (REH) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.