Learn More
Eurofins Discoverx PathHunter Human CD200R Signaling Cell-Based Assay Kit (Jurkat)
Catalog No. 5000456169
Encompass
The PathHunter Human CD200R (DOK2) Signaling Stable Cell Line Assay (Jurkat) measures CD200R signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56169 Supplier Eurofins Discoverx Supplier No. 7931176C19
Add to Cart
Add to Cart
The PathHunter Human CD200R (DOK2) Signaling Stable Cell Line Assay (Jurkat) measures CD200R signal by recruitment of SH2 domain protein to the activated receptor. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.