Learn More
Eurofins Discoverx PathHunter Human c-MET/EGFR Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455463
Encompass
The PathHunter Human c-MET/EGFR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of c-Met/EGFR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55463 Supplier Eurofins Discoverx Supplier No. 7931179C3
Add to Cart
Add to Cart
The PathHunter Human c-MET/EGFR Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of c-Met/EGFR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.