Learn More
Eurofins Discoverx PathHunter Human TSLP Signaling Reporter Cell-Based Assay Kit (REH)
Catalog No. 5000456143
Encompass
The PathHunter Human TSLP Signaling STAT5 Reporter Stable Cell Line Assay (REH) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56143 Supplier Eurofins Discoverx Supplier No. 7931182C055
The PathHunter Human TSLP Signaling STAT5 Reporter Stable Cell Line Assay (REH) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.