Learn More
Eurofins Discoverx PathHunter Human APRIL (BCMA) Signaling Reporter Cell-Based Assay Kit (K562)
Catalog No. 5000455759
Encompass
The PathHunter Human APRIL (BCMA) Signaling NF-kB Reporter Stable Cell Line Assay (K562) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55759 Supplier Eurofins Discoverx Supplier No. 7931183C042
The PathHunter Human APRIL (BCMA) Signaling NF-kB Reporter Stable Cell Line Assay (K562) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC). This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.