Learn More
Eurofins Discoverx PathHunter Human LPS Signaling Reporter Cell-Based Assay Kit (THP-1)
Catalog No. 5000455500
Encompass
The PathHunter Human LPS Signaling NF-kB Reporter Stable Cell Line Assay (THP-1) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55500 Supplier Eurofins Discoverx Supplier No. 7931187C14
The PathHunter Human LPS Signaling NF-kB Reporter Stable Cell Line Assay (THP-1) monitor the activation of specific signaling pathways, utilizing the established PathHunter Enzyme Fragment Complementation (EFC) technology. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.