Learn More
Eurofins Discoverx PathHunter Human MC2R (ACTH) β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000455856
Encompass
The PathHunter Human MC2R (ACTHR) β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure MC2R (GPCR) activity via recruitment of β-Arrestin 2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55856 Supplier Eurofins Discoverx Supplier No. 7931189C3
The PathHunter Human MC2R (ACTHR) β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure MC2R (GPCR) activity via recruitment of β-Arrestin 2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.