Learn More
Eurofins Discoverx cAMP Hunter™ Human GPBAR1 Gs Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456089
Encompass
The cAMP Hunter™ Human GPBAR1 Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GPBAR1 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56089 Supplier Eurofins Discoverx Supplier No. 7950049C2
The cAMP Hunter™ Human GPBAR1 Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GPBAR1 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.