Learn More
Eurofins Discoverx cAMP Hunter™ Human PTHR2 Gs/Gq Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455910
Encompass
The cAMP Hunter™ Human PTHR2 Gs/Gq Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure PTHR2 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55910 Supplier Eurofins Discoverx Supplier No. 7950057C2
Add to Cart
Add to Cart
The cAMP Hunter™ Human PTHR2 Gs/Gq Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure PTHR2 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.