Learn More
Eurofins Discoverx cAMP Hunter™ Human GLP-1 (GLP-1 Receptor) Gs Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456040
Encompass
The cAMP Hunter™ Human GLP-1 (GLP1R) Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56040 Supplier Eurofins Discoverx Supplier No. 7950062C2
The cAMP Hunter™ Human GLP-1 (GLP1R) Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.