Learn More
Eurofins Discoverx PathHunter Human SDF-1 alpha (CXCR7) β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455633
Encompass
The PathHunter Human SDF-1 alpha (CXCR7) β-Arrestin Stable Cell Line Assay (CHO-K1) CXCR7 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55633 Supplier Eurofins Discoverx Supplier No. 7930248C2
The PathHunter Human SDF-1 alpha (CXCR7) β-Arrestin Stable Cell Line Assay (CHO-K1) CXCR7 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.