Learn More
Eurofins Discoverx cAMP Hunter™ Human OPRD1 (Opioid delta) Gi Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455929
Encompass
The cAMP Hunter™ Human OPRD1 (Opioid delta) Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure OPRD1 (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55929 Supplier Eurofins Discoverx Supplier No. 7950108C2
The cAMP Hunter™ Human OPRD1 (Opioid delta) Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure OPRD1 (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.