Learn More
Eurofins Discoverx cAMP Hunter™ Human MC1R Gs Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7950111C2
Encompass
The cAMP Hunter™ Human MC1R Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure MC1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55511