Learn More
Eurofins Discoverx cAMP Hunter™ Rat DRD2S Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456011
Encompass
The cAMP Hunter™ Rat DRD2S Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure rat DRD2S (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56011 Supplier Eurofins Discoverx Supplier No. 7950129C2
The cAMP Hunter™ Rat DRD2S Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure rat DRD2S (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.