Learn More
Eurofins Discoverx cAMP Hunter™ Human RXFP4 Gi Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455959
Encompass
The cAMP Hunter™ Human RXFP4 Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure RXFP4 (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55959 Supplier Eurofins Discoverx Supplier No. 7950134C2
The cAMP Hunter™ Human RXFP4 Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure RXFP4 (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.